Learn More
Biomatik Corporation Human Collagen Alpha-1 (Xvii) Chain (Col17A1) Elisa Kit
Supplier:  Biomatik Corporation EKC33206
Encompass_Preferred
Human Collagen Alpha-1 (Xvii) Chain (Col17A1) Elisa Kit for detecting Collagen, Type Xvii, Alpha 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9UMD9; COL17A1. Target Synonyms: COL17A1; BP180; BPAG2Collagen alpha-1 (XVII) chain; 180 kDa bullous pemphigoid antigen 2; Bullous pemphigoid antigen 2) [Cleaved into: 120 kDa linear IgA disease antigen; 120 kDa linear IgA dermatosis antigen; Linear IgA disease antigen 1; LAD-1); 97 kDa linear IgA disease antigen; 97 kDa linear IgA bullous dermatosis antigen; 97 kDa LAD antigen; 97-LAD; Linear IgA bullous disease antigen of 97 kDa; LABD97)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3879