Learn More
Biomatik Corporation Human Collagen Alpha-3 (Vi) Chain (Col6A3) Elisa Kit
Supplier:  Biomatik Corporation EKC33217
Encompass_Preferred
Human Collagen Alpha-3 (Vi) Chain (Col6A3) Elisa Kit for detecting Collagen, Type Vi, Alpha 3 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.39ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P12111; COL6A3. Target Synonyms: CO6A3_HUMAN; COL6A3; Collagen alpha-3 (VI) chain; Collagen type VI alpha 3; Collagen VI alpha 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3881