Learn More
Biomatik Corporation Human Complement Component C8 Alpha Chain (C8A) Elisa Kit
Supplier:  Biomatik Corporation EKC33262
Encompass_Preferred
Human Complement Component C8 Alpha Chain (C8A) Elisa Kit for detecting Complement Component 8, Alpha Polypeptide in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.39ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P07357; C8A. Target Synonyms: C8A; CO8A_HUMAN; Complement component 8 subunit alpha; Complement component 8; alpha polypeptide; Complement component C8 alpha chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3890