Learn More
Biomatik Corporation Human Complement Factor H-Related Protein 1 (Cfhr1) Elisa Kit
Supplier:  Biomatik Corporation EKC33266
Encompass_Preferred
Human Complement Factor H-Related Protein 1 (Cfhr1) Elisa Kit for detecting Complement Factor H-Related 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 25-1600ng/ml. Sensitivity: 12.5ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q03591; CFHR1. Target Synonyms: CFHL; CFHL1; CFHL1P; CFHR1; CFHR1P; Complement factor H related 1; Complement factor H related 1 pseudogene; Complement factor H related protein 1 precursor; Complement factor H-related protein 1; FHR 1; FHR-1; FHR1; FHR1_HUMAN; H factor (complement) like 1; H factor (complement) like 2; H factor like protein 1; H factor-like protein 1; H-factor-like 1; H36 1; H36 2; H36; HFL1; HFL2; MGC104329. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3892