Learn More
Biomatik Corporation Human Complement fragment 3b,C3b ELISA Kit
Supplier:  Biomatik Corporation EKC33274
Encompass_Preferred
Sample Type: serum, urine, cerebrospinal fluid (CSF), saliva. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12 ng/mL-200 ng/mL. Sensitivity: 0.78 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-3894