Learn More
Biomatik Corporation Human DNA damage-inducible transcript 4 protein(DDIT4) ELISA kit
Supplier:  Biomatik Corporation EKC33482
Encompass_Preferred
Sample Type: serum, tissue homogenates, cell lysates, urine, cerebrospinal fluid (CSF). Species Reactivity: Homo sapiens (Human). Detection Range: 0.156 ng/mL-10 ng/mL. Sensitivity: 0.039 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9NX09. Target Synonyms: Dig2, FLJ20500, REDD-1, REDD1, RP11-442H21.1, RTP801, HIF-1 responsive RTP801 (RTP801)|RTP801. Restriction: For Research Use Only.
Catalog No. 50-229-3930