Learn More
Biomatik Corporation Human enkephalin,ENK ELISA Kit
Supplier:  Biomatik Corporation EKC33575
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 0.05 ng/mL-10 ng/mL. Sensitivity: 0.025 ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-3945