Learn More
Biomatik Corporation Human Ephrin Type-B Receptor 2 (Ephb2) Elisa Kit
Supplier:  Biomatik Corporation EKC33587
Encompass_Preferred
Human Ephrin Type-B Receptor 2 (Ephb2) Elisa Kit for detecting Eph Receptor B2 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P29323; EPHB2. Target Synonyms: cek5; Developmentally regulated EPH related tyrosine kinase; DRT; EK5; ELK related protein tyrosine kinase; Eph receptor B2; EPH tyrosine kinase 3; EPH-like kinase 5; EPHB2; EPHB2_HUMAN; Ephrin type B receptor 2; Ephrin type-B receptor 2; EPHT 3; ERK; ETECK; hEK5; Nuk; Prkm 5; Receptor protein tyrosine kinase HEK 5; Renal carcinoma antigen NY-REN-47; Sek 3; Tyro 5; Tyrosine protein kinase receptor CEK 5; Tyrosine protein kinase receptor EPH 3; Tyrosine protein kinase receptor QEK 5; Tyrosine-protein kinase receptor EPH-3; Tyrosine-protein kinase TYRO5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3950