Learn More
Biomatik Corporation Human Erbb Receptor Feedback Inhibitor 1 (Errfi1) Elisa Kit
Supplier:  Biomatik Corporation EKC33627
Encompass_Preferred
Human Erbb Receptor Feedback Inhibitor 1 (Errfi1) Elisa Kit for detecting Erbb Receptor Feedback Inhibitor 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9UJM3; ERRFI1. Target Synonyms: ERBB receptor feedback inhibitor 1; ERRFI_HUMAN; Errfi1; GENE 33; Gene 33; rat; homolog of; Mig-6; MIG6; mitogen inducible gene 6; mitogen inducible gene 6 protein; Mitogen-inducible gene 6 protein; RALT; receptor-associated late transducer. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3954