Learn More
Biomatik Corporation Human F-Box Only Protein 32 (Fbxo32) Elisa Kit
Supplier:  Biomatik Corporation EKC33669
Encompass_Preferred
Human F-Box Only Protein 32 (Fbxo32) Elisa Kit for detecting F-Box Protein 32 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q969P5; FBXO32. Target Synonyms: 4833442G10Rik; AI430017; Atrogin 1; Atrogin-1; ATROGIN1; Atrophy gene 1; F box only protein 32; F-box only protein 32; F-box protein 32; FBX32_HUMAN; fbxo25; FBXO32; FLJ32424; MAFbx; MGC108443; MGC137646; MGC33610; Muscle atrophy F box; Muscle atrophy F box protein; Muscle atrophy F-box protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3960