Learn More
Biomatik Corporation Human Fibrinogen Alpha Chain (Fga) Elisa Kit
Supplier:  Biomatik Corporation EKC33681
Encompass_Preferred
Human Fibrinogen Alpha Chain (Fga) Elisa Kit for detecting Fibrinogen Alpha Chain (Fga) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.94-60ug/ml. Sensitivity: 0.47ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P02671; FGA. Target Synonyms: FGA; FGB; FGG; Fib2; FIBA_HUMAN; Fibrinogen A alpha polypeptide; Fibrinogen alpha chain; Fibrinogen B alpha polypeptide; Fibrinogen beta chain; Fibrinogen G alpha polypeptide; Fibrinogen gamma chain; fibrinogen; B beta polypeptide; fibrinogen; G gamma polypeptide; fibrinogen; gamma polypeptide; Fibrinogen--alpha -polypeptide chain; Fibrinogen--beta -polypeptide chain; Fibrinogen--gamma-polypeptide chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3964