Learn More
Biomatik Corporation Human Fibroblast Growth Factor 10 (Fgf10) Elisa Kit
Supplier:  Biomatik Corporation EKC33688
Encompass_Preferred
Human Fibroblast Growth Factor 10 (Fgf10) Elisa Kit for detecting Fibroblast Growth Factor 10 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200pg/ml. Sensitivity: 0.78pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O15520; FGF10. Target Synonyms: BB213776; fd11d03; FGF 10; FGF-10; FGF10; FGF10_HUMAN; Fibroblast growth factor 10; Keratinocyte growth factor 2; KGF 2; Produced by fibroblasts of urinary bladder lamina propria; wu:fd11d03; zgc:109774. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3966