Learn More
Biomatik Corporation Human Flotillin-1 (Flot1) Elisa Kit
Supplier:  Biomatik Corporation EKC33731
Encompass_Preferred
Human Flotillin-1 (Flot1) Elisa Kit for detecting Flotillin 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 12.5-800pg/ml. Sensitivity: 3.12pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O75955; FLOT1. Target Synonyms: FLOT 1; FLOT1; FLOT1_HUMAN; Flotillin-1; Flotillin1; Integral membrane component of caveolae; Reggie 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3972