Learn More
Biomatik Corporation Human Fragile X Mental Retardation 1 Protein (Fmr1) Elisa Kit
Supplier:  Biomatik Corporation EKC33753
Encompass_Preferred
Human Fragile X Mental Retardation 1 Protein (Fmr1) Elisa Kit for detecting Fragile X Mental Retardation 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q06787; FMR1. Target Synonyms: FMR 1; Fmr1; Fmr1 gene; FMR1_HUMAN; FMRP; Fragile X mental retardation 1; Fragile X mental retardation 1 protein; Fragile X mental retardation protein 1; Fragile X mental retardation protein; fragile X mental retardation syndrome-related protein 1; fragile X mental retardation; autosomal homolog 1; FRAXA; fxr1; MGC87458; POF; POF1; Protein FMR-1; Protein FMR1; wu:fb16f11; wu:fd18c10; zgc:66226. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3978