Learn More
Biomatik Corporation Human Glutathione Peroxidase 1 (Gpx1) Elisa Kit
Supplier:  Biomatik Corporation EKC33852
Encompass_Preferred
Human Glutathione Peroxidase 1 (Gpx1) Elisa Kit for detecting Glutathione Peroxidase 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000uU/ml. Sensitivity: 3.9uU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P07203; GPX1. Target Synonyms: AL033363; Cellular glutathione peroxidase; Glutathione peroxidase 1; Glutathione peroxidase; GPx 1; GPx-1; GPX1; GPX1_HUMAN; GPXD; GSHPx-1; GSHPX1; MGC14399; MGC88245. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3997