Learn More
Biomatik Corporation Human Glutathione Reductase (Gr) Elisa Kit
Supplier:  Biomatik Corporation EKC33857
Encompass_Preferred
Human Glutathione Reductase (Gr) Elisa Kit for detecting Glutathione Reductase in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40mU/ml. Sensitivity: 0.156mU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P00390; GSR. Target Synonyms: GLR1; GLUR; Glutathione reductase; Glutathione reductase mitochondrial; Glutathione reductase; mitochondrial; GR; Gr1; GRase; GRD 1; GRD1; GSHR_HUMAN; GSR; MGC78522. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3999