Learn More
Biomatik Corporation Human Glycodelin Elisa Kit
Supplier:  Biomatik Corporation EKC33869
Encompass_Preferred
Human Glycodelin Elisa Kit for detecting Progestagen-Associated Endometrial Protein in serum, cerebrospinal fluid (csf), urine, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.195ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P09466; PAEP. Target Synonyms: Alpha uterine protein; gD; GdA; GdF; GdS; Glycodelin A; Glycodelin; Glycodelin F; Glycodelin S; MGC138509; MGC142288; PAEG; PAEP; PAEP_HUMAN; PEG; PEP; Placental Protein 14 / Glycodelin A; Placental protein 14; PP14; Pregnancy-associated endometrial alpha-2 globulin; Progestagen-associated endometrial protein; Progesterone-associated endometrial protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4003