Learn More
Biomatik Corporation Human Gonadotropin-Releasing Hormone, Gnrh Elisa Kit
Supplier:  Biomatik Corporation EKC33888
Encompass_Preferred
Human Gonadotropin-Releasing Hormone, Gnrh Elisa Kit for detecting Gonadotropin-Releasing Hormone 1 (Luteinizing-Releasing Hormone) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 2.5-100pg/ml. Sensitivity: 0.625pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01148; GNRH1. Target Synonyms: GNRH1; GNRH; GRH; LHRHProgonadoliberin-1; Progonadoliberin I) [Cleaved into: Gonadoliberin-1; Gonadoliberin I; Gonadorelin; Gonadotropin-releasing hormone I; GnRH-I; Luliberin I; Luteinizing hormone-releasing hormone I; LH-RH I); GnRH-associated peptide 1; GnRH-associated peptide I)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4009