Learn More
Biomatik Corporation Human Group IIF secretory phospholipase A2(PLA2G2F) ELISA kit
Supplier:  Biomatik Corporation EKC33908
Encompass_Preferred
Sample Type: serum, plasma, tissue,homogenates. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312 ng/mL-20 ng/mL. Sensitivity: 0.078 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9BZM2. Target Synonyms: RP3-340N1.1, FLJ25429, FLJ36326. Restriction: For Research Use Only.
Catalog No. 50-229-4013