Learn More
Biomatik Corporation Human Growth Arrest And Dna Damage-Inducible Protein Gadd45 Alpha (Gadd45A) Elisa Kit
Supplier:  Biomatik Corporation EKC33910
Encompass_Preferred
Human Growth Arrest And Dna Damage-Inducible Protein Gadd45 Alpha (Gadd45A) Elisa Kit for detecting Growth Arrest And Dna-Damage-Inducible, Alpha in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 25-1600pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P24522; GADD45A. Target Synonyms: DDIT 1; DDIT-1; DDIT1; DNA damage inducible transcript 1; DNA damage-inducible transcript 1 protein; GA45A_HUMAN; GADD45; GADD45A; Growth arrest and DNA damage inducible 45 alpha; Growth arrest and DNA damage inducible alpha; Growth arrest and DNA damage-inducible protein GADD45 alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4015