Learn More
Biomatik Corporation Human H-Cadherin, H-Cad/Cdh13 Elisa Kit
Supplier:  Biomatik Corporation EKC33941
Encompass_Preferred
Human H-Cadherin, H-Cad/Cdh13 Elisa Kit for detecting Cadherin 13, H-Cadherin (Heart) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.25-80ng/ml. Sensitivity: 0.312ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P55290; CDH13. Target Synonyms: CAD13_HUMAN; Cadherin-13; CDH13; CDHH; H-cadherin; Heart cadherin; P105; T cad; T Cadherin; T-cad; T-cadherin; Truncated cadherin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4018