Learn More
Biomatik Corporation Human Hemoglobin Subunit Delta (Hbd) Elisa Kit
Supplier:  Biomatik Corporation EKC33972
Encompass_Preferred
Human Hemoglobin Subunit Delta (Hbd) Elisa Kit for detecting Hemoglobin, Delta in serum, plasma, lysate for rbc. Species Reactivity: Human (Homo sapiens). Detection Range: 0.007-30ng/ml. Sensitivity: 0.007ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P02042; HBD. Target Synonyms: Delta-globin; HBD; HBD_HUMAN; Hemoglobin delta chain; Hemoglobin subunit delta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4025