Learn More
Biomatik Corporation Human Heparin Cofactor Ii, Hciielisa Kit
Supplier:  Biomatik Corporation EKC33987
Encompass_Preferred
Human Heparin Cofactor Ii, Hciielisa Kit for detecting Serpin Peptidase Inhibitor, Clade D (Heparin Cofactor), Member 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.25-80pg/ml. Sensitivity: 0.312pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P05546; SERPIND1. Target Synonyms: D22S673; HC II; HC-II; HC2; HCF2; HEP2_HUMAN; Heparin cofactor 2; Heparin cofactor II; HLS2; Leuserpin 2; LS2; Protease inhibitor leuserpin 2; Protease inhibitor leuserpin-2; serine (or cysteine) proteinase inhibitor; clade D (heparin cofactor); member 1; Serpin D1; serpin peptidase inhibitor; clade D (heparin cofactor); member 1; SERPIND1; THPH10. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4027