Learn More
Biomatik Corporation Human High Molecular Weight Adiponectin (Hmw Adiponectin) Elisa Kit
Supplier:  Biomatik Corporation EKC34029
Encompass_Preferred
Human High Molecular Weight Adiponectin (Hmw Adiponectin) Elisa Kit for detecting High Molecular Weight Adiponectin(Hmw Adiponectin) in serum, plasma, cell culture supernates, tissue homogenates, urine. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 0.78ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q15848; ADIPOQ. Target Synonyms: 30 kDa adipocyte complement related protein; 30 kDa adipocyte complement-related protein; ACDC; Acrp 30; ACRP30; ADIPO_HUMAN; Adipocyte; Adipocyte C1q and collagen domain containing protein; Adipocyte complement related 30 kDa protein; Adipocyte complement related protein of 30 kDa; Adipocyte complement-related 30 kDa protein; adipocyte-specific secretory protein; Adiponectin; Adiponectin precursor; adiponectin; C1Q and collagen domain containing. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4029