Learn More
Biomatik Corporation Human Hyaluronan-Binding Protein 2 (Habp2) Elisa Kit
Supplier:  Biomatik Corporation EKC34083
Encompass_Preferred
Human Hyaluronan-Binding Protein 2 (Habp2) Elisa Kit for detecting Hyaluronan Binding Protein 2 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q14520; HABP2. Target Synonyms: Factor seven activating protease; Factor seven-activating protease; Factor VII activating protein; Factor VII-activating protease; FSAP; HABP 2; HABP; Habp2; HABP2_HUMAN; Hepatocyte growth factor activator like protein; Hepatocyte growth factor activator-like protein; HGFAL; Hyaluronan binding protein 2; Hyaluronan-binding protein 2 27 kDa light chain alternate form; Hyaluronic acid binding protein 2; PHBP; Plasma hyaluronan binding protein; Plasma hyaluronan-binding protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4038