Learn More
Biomatik Corporation Human Hyaluronidase-2 (Hyal2) Elisa Kit
Supplier:  Biomatik Corporation EKC34087
Encompass_Preferred
Human Hyaluronidase-2 (Hyal2) Elisa Kit for detecting Hyaluronoglucosaminidase 2 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q12891; HYAL2. Target Synonyms: Hyal 2; Hyal-2; Hyal2; HYAL2_HUMAN; Hyaluronidase 2; Hyaluronidase 2 Precursor; Hyaluronidase-2; Hyaluronoglucosaminidase 2; Hyaluronoglucosaminidase-2; LUCA 2; LUCA-2; LUCA2; Lung carcinoma protein 2; lysosomal hyaluronidase; PH 20 homolog; PH-20 homolog; PH20 homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4040