Learn More
Biomatik Corporation Human Integrin Beta-1 (Itgb1) Elisa Kit
Supplier:  Biomatik Corporation EKC34169
Encompass_Preferred
Human Integrin Beta-1 (Itgb1) Elisa Kit for detecting Integrin, Beta 1 (Fibronectin Receptor, Beta Polypeptide, Antigen Cd29 Includes Mdf2, Msk12) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P05556; ITGB1. Target Synonyms: ITGB1; FNRB; MDF2; MSK12; Integrin beta-1; Fibronectin receptor subunit beta; Glycoprotein IIa; GPIIA; VLA-4 subunit beta; CD antigen CD29. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4056