Learn More
Biomatik Corporation Human Inter-Alpha-Trypsin Inhibitor Heavy Chain H3 (Itih3) Elisa Kit
Supplier:  Biomatik Corporation EKC34182
Encompass_Preferred
Human Inter-Alpha-Trypsin Inhibitor Heavy Chain H3 (Itih3) Elisa Kit for detecting Inter-Alpha (Globulin) Inhibitor H3 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 78.13-5000ng/ml. Sensitivity: 19.53pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q06033; ITIH3. Target Synonyms: ITIH3; Inter-alpha-trypsin inhibitor heavy chain H3; ITI heavy chain H3; ITI-HC3; Inter-alpha-inhibitor heavy chain 3; Serum-derived hyaluronan-associated protein; SHAP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4059