Learn More
Biomatik Corporation Human Interferon Alpha-21 (Ifna21) Elisa Kit
Supplier:  Biomatik Corporation EKC34193
Encompass_Preferred
Human Interferon Alpha-21 (Ifna21) Elisa Kit for detecting Interferon, Alpha 21 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01568; IFNA21. Target Synonyms: IFNA21; Interferon alpha-21; IFN-alpha-21; Interferon alpha-F; LeIF F. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4061