Learn More
Biomatik Corporation Human Interleukin-36 Gamma (Il1F9) Elisa Kit
Supplier:  Biomatik Corporation EKC34301
Encompass_Preferred
Human Interleukin-36 Gamma (Il1F9) Elisa Kit for detecting Interleukin 1 Family, Member 9 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9NZH8; IL36G. Target Synonyms: IL 1 epsilon; IL 1 related protein 2; IL 1 (EPSILON); IL 1F9; IL 1H1; IL 1RP2; IL-1 epsilon; IL-1-related protein 2; IL-1F9; IL-1H1; IL-1RP2; IL1E; Il1f9; IL1F9_HUMAN; IL1H1; IL1RP2; IL36 gamma; IL36G; Interleukin 1 epsilon; Interleukin 1 family member 9; Interleukin 1 homolog 1; Interleukin 1 related protein 2; Interleukin 36 gamma; Interleukin-1 epsilon; Interleukin-1 family member 9; Interleukin-1 homolog 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4076