Learn More
Biomatik Corporation Human Keratin, Type I Cytoskeletal 23 (Krt23) Elisa Kit
Supplier:  Biomatik Corporation EKC34336
Encompass_Preferred
Human Keratin, Type I Cytoskeletal 23 (Krt23) Elisa Kit for detecting Keratin 23 (Histone Deacetylase Inducible) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.9-60ng/ml. Sensitivity: 0.23ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9C075; KRT23. Target Synonyms: CK 23; CK-23; CK23; Cytokeratin 23; Cytokeratin-23; Cytokeratin23; DKFZP434G032; HAIK 1; HAIK1; Histone deacetylase inducible keratin 23; Hyperacetylation inducible type I keratin; K1C23_HUMAN; K23; Keratin 23 (histone deacetylase inducible); Keratin 23; Keratin; Keratin type I cytoskeletal 23; Keratin-23; Keratin23; KRT 23; KRT23; MGC26158; type I cytoskeletal 23; Type I intermediate filament cytokeratin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4081