Learn More
Biomatik Corporation Mouse Epithelial neutrophil activating peptide 78,ENA-78 ELISA Kit
Supplier:  Biomatik Corporation EKC42253
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 62.5 pg/ml - 4000 pg/ml. Sensitivity: 15.6 pg/ml. Format: 96-Well Plate. Assay Type: Not Available. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P50228. Target Synonyms: ENA-78, SCYB5, epithelial-derived neutrophil activating protein 78|epithelial-derived neutrophil-activating peptide 78|neutrophil-activating peptide ENA-78|neutrophil-activating protein 78|small ind. Restriction: For Research Use Only.
Catalog No. 50-229-5027