Learn More
Biomatik Corporation Rat soluble Cluster of differentiation 86,sCD86 ELISA Kit
Supplier:  Biomatik Corporation EKC42266
Encompass_Preferred
Sample Type: serum, plasma,tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 78 pg/mL-5000 pg/mL. Sensitivity: 19.5 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: O35531. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-5032