Learn More
Biomatik Corporation Mouse granzyme A (GZMA) ELISA Kit
Supplier:  Biomatik Corporation EKC42272
Encompass_Preferred
Sample Type: serum, plasma,tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 6.25 pg/mL-400 pg/mL. Sensitivity: 1.56 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P11032. Target Synonyms: CTLA3, HFSP, CTL tryptase|Granzyme A (Cytotoxic T-lymphocyte-associated serine esterase-3; Hanukah factor serine protease)|granzyme A. Restriction: For Research Use Only.
Catalog No. 50-229-5034