Learn More
Biomatik Corporation Bovine folic acid,FA ELISA Kit
Supplier:  Biomatik Corporation EKC42307
Encompass_Preferred
Sample Type: serum, plasma,tissue homogenates. Species Reactivity: Bos taurus (Bovine). Detection Range: 0.39 ng/mL-100 ng/mL. Sensitivity: 0.39 ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-5046