Learn More
Biomatik Corporation Human Α-Subunits Of Hemoglobin (Hb-Α) Elisa Kit
Supplier:  Biomatik Corporation EKC42311
Encompass_Preferred
Human Α-Subunits Of Hemoglobin (Hb-Α) Elisa Kit for detecting Hemoglobin, Alpha 2 in serum, plasma, lysate for rbc. Species Reactivity: Human (Homo sapiens). Detection Range: 0.170-700ng/ml. Sensitivity: 0.17ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P69905; HBA1. Target Synonyms: Alpha 1 globin; Alpha globin; Alpha one globin; Alpha-globin; HBA_HUMAN; HBA1; HBA2; Hemoglobin alpha 1; Hemoglobin alpha 1 chain; Hemoglobin alpha 1 globin chain; Hemoglobin alpha 2; Hemoglobin alpha chain; Hemoglobin subunit alpha; MGC126895; MGC126897. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5048