Learn More
Biomatik Corporation Human Soluble Cd23 (Scd23) Elisa Kit
Supplier:  Biomatik Corporation EKC42330
Encompass_Preferred
Human Soluble Cd23 (Scd23) Elisa Kit for detecting Soluble Cd23(Scd23) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P06734; FCER2. Target Synonyms: Blast 2; BLAST-2; Blast2; C type lectin domain family 4 member J; C-type lectin domain family 4; C-type lectin domain family 4 member J; C-type lectin domain family 4; member J; CD 23; CD 23A; CD23; CD23 antigen; CD23A; CLEC 4J; CLEC4J; Fc epsilon receptor II; Fc epsilon RII; Fc fragment of IgE; Fc fragment of IgE low affinity II receptor for; Fc fragment of IgE receptor II; Fc fragment of IgE; low affinity II; receptor for (CD23); Fc of IgE; Fc of IgE; low affinity II; receptor for (CD23). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5053