Learn More
Biomatik Corporation Fish hepatocyte growth factor(HGF)ELISA Kit
Supplier:  Biomatik Corporation EKC42344
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Fish. Detection Range: 200 pg/mL-8000 pg/mL. Sensitivity: 167 pg/mL. Format: 96-Well Plate. Assay Type: Not Available. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: F1REK1. Target Synonyms: DFNB39, F-TCF, HGFB, HPTA, SF, OTTHUMP00000206710|fibroblast-derived tumor cytotoxic factor|hepatocyte growth factor|lung fibroblast-derived mitogen. Restriction: For Research Use Only.
Catalog No. 50-229-5057