Learn More
Biomatik Corporation Human Disintegrin And Metalloproteinase Domain-Containing Protein 2 (Adam2) Elisa Kit
Supplier:  Biomatik Corporation EKC42362
Encompass_Preferred
Human Disintegrin And Metalloproteinase Domain-Containing Protein 2 (Adam2) Elisa Kit for detecting Adam Metallopeptidase Domain 2 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q99965; ADAM2. Target Synonyms: A disintegrin and metalloproteinase domain 2; ADAM 2; ADAM metallopeptidase domain 2; Adam2; ADAM2_HUMAN; AI323749; Cancer/testis antigen 15; CRYN1; CRYN2; CT15; Disintegrin and metalloproteinase domain-containing protein 2; Fertilin beta; Fertilin subunit beta; Ftnb; PH 30; PH 30b; PH-30; PH30; Ph30 beta; PH30-beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5059