Learn More
Biomatik Corporation Human Natural Killer Cell Receptor 2B4 (Cd244) Elisa Kit
Supplier:  Biomatik Corporation EKC42411
Encompass_Preferred
Human Natural Killer Cell Receptor 2B4 (Cd244) Elisa Kit for detecting Cd244 Molecule, Natural Killer Cell Receptor 2B4 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9BZW8; CD244. Target Synonyms: 2B4; C9.1; CD244; CD244 antigen; CD244 molecule; CD244 molecule natural killer cell receptor 2B4; CD244 natural killer cell receptor 2B4; CD244_HUMAN; F730046O15Rik; h2B4; Ly90; NAIL; Natural killer cell activation-inducing ligand; Natural killer cell receptor 2B4; NK cell activation inducing ligand; NK cell activation inducing ligand NAIL; NK cell activation-inducing ligand; NK cell type I receptor protein 2B4; NKR2B4; Nmrk; Non-MHC restricted killing associated; OTTHUMP00000027884; p38. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5075