Learn More
Biomatik Corporation Human Adenosine Deaminase Cecr1 (Cecr1) Elisa Kit
Supplier:  Biomatik Corporation EKC42427
Encompass_Preferred
Human Adenosine Deaminase Cecr1 (Cecr1) Elisa Kit for detecting Cat Eye Syndrome Chromosome Region, Candidate 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9NZK5; ADA2. Target Synonyms: ADA2; ADGF; CECR1; IDGFLAdenosine deaminase 2; EC 3.5.4.4; Cat eye syndrome critical region protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5078