Learn More
Biomatik Corporation Human Carboxypeptidase N Subunit 2 (Cpn2) Elisa Kit
Supplier:  Biomatik Corporation EKC42444
Encompass_Preferred
Human Carboxypeptidase N Subunit 2 (Cpn2) Elisa Kit for detecting Carboxypeptidase N, Polypeptide 2 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P22792; CPN2. Target Synonyms: CPN2; ACBPCarboxypeptidase N subunit 2; Carboxypeptidase N 83 kDa chain; Carboxypeptidase N large subunit; Carboxypeptidase N polypeptide 2; Carboxypeptidase N regulatory subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5081