Learn More
Biomatik Corporation Human Nadh-Cytochrome B5 Reductase 3 (Cyb5R3) Elisa Kit
Supplier:  Biomatik Corporation EKC42457
Encompass_Preferred
Human Nadh-Cytochrome B5 Reductase 3 (Cyb5R3) Elisa Kit for detecting Cytochrome B5 Reductase 3 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 25-1600pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P00387; CYB5R3. Target Synonyms: B5R; Cyb5r3; Cytochrome b5 reductase 3; Cytochrome b5 reductase; DIA1; Diaphorase 1; Diaphorase-1; NADH cytochrome b5 reductase 3; NADH-cytochrome b5 reductase 3 membrane-bound form; NADH-cytochrome b5 reductase 3 soluble form; NB5R3_HUMAN; OTTHUMP00000028761; OTTHUMP00000198435; OTTHUMP00000198574; OTTHUMP00000198662; OTTHUMP00000198665. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5085