Learn More
Biomatik Corporation Human Fatty Acid-Binding Protein 12 (Fabp12) Elisa Kit
Supplier:  Biomatik Corporation EKC42477
Encompass_Preferred
Human Fatty Acid-Binding Protein 12 (Fabp12) Elisa Kit for detecting Fatty Acid Binding Protein 12 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: A6NFH5; FABP12. Target Synonyms: FABP12; Fatty acid binding protein ORF; Fatty acid-binding protein 12; FBP12_HUMAN; Intracellular fatty acid binding protein FABP12; Probable fatty acid binding protein ENSP00000353650. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5094