Learn More
Biomatik Corporation Mouse Ficolin-2(FCN2) ELISA kit
Supplier:  Biomatik Corporation EKC42481
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 125 pg/mL-8000 pg/mL. Sensitivity: 31.25 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: O70497. Target Synonyms: RP11-263F14.2, EBP-37, FCNL, P35, ficolin-2, L-ficolin|collagen/fibrinogen domain-containing protein 2|ficolin 2|ficolin B|serum lectin p35. Restriction: For Research Use Only.
Catalog No. 50-229-5096