Learn More
Biomatik Corporation Human Gamma-Aminobutyric Acid Receptor-Associated Protein (Gabarap) Elisa Kit
Supplier:  Biomatik Corporation EKC42492
Encompass_Preferred
Human Gamma-Aminobutyric Acid Receptor-Associated Protein (Gabarap) Elisa Kit for detecting Gaba(A) Receptor-Associated Protein in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 39.06-2500pg/ml. Sensitivity: 9.77pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O95166; GABARAP. Target Synonyms: ATG8A; FLC 3B; FLC3B; FLJ25768; GABA type A receptor associated protein; GABA (A) receptor associated protein; GABA (A) receptor-associated protein; GABARAP a; GABARAP; Gamma aminobutyric acid receptor associated protein; Gamma-aminobutyric acid receptor-associated protein; GBRAP_HUMAN; MGC120154; MGC120155; MM 46; MM46. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5101