Learn More
Biomatik Corporation Human Transmembrane Glycoprotein Nmb (Gpnmb) Elisa Kit
Supplier:  Biomatik Corporation EKC42499
Encompass_Preferred
Human Transmembrane Glycoprotein Nmb (Gpnmb) Elisa Kit for detecting Glycoprotein (Transmembrane) Nmb in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.195ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q14956; GPNMB. Target Synonyms: Glycoprotein (transmembrane) nmb; Glycoprotein nmb; Glycoprotein nmb like protein; Gpnmb; GPNMB_HUMAN; HGFIN; NMB; Osteoactivin; Transmembrane glycoprotein; Transmembrane glycoprotein HGFIN; Transmembrane glycoprotein NMB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5103