Learn More
Biomatik Corporation Mouse Apoptosis-Associated Speck-Like Protein Containing A Card (Pycard) Elisa Kit
Supplier:  Biomatik Corporation EKC42603
Encompass_Preferred
Mouse Apoptosis-Associated Speck-Like Protein Containing A Card (Pycard) Elisa Kit for detecting Pyd And Card Domain Containing in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.81-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9EPB4; Pycard. Target Synonyms: Pycard; AscApoptosis-associated speck-like protein containing a CARD; mASC; PYD and CARD domain-containing protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5138