Learn More
Biomatik Corporation Human Kallistatin (Serpina4) Elisa Kit
Supplier:  Biomatik Corporation EKC42621
Encompass_Preferred
Human Kallistatin (Serpina4) Elisa Kit for detecting Serpin Peptidase Inhibitor, Clade A (Alpha-1 Antiproteinase, Antitrypsin), Member 4 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P29622; SERPINA4. Target Synonyms: KAIN_HUMAN; KAL; Kallikrein binding protein; Kallikrein inhibitor; Kallistatin; KBP; KLST; KST; MGC108582; Peptidase inhibitor 4; PI-4; PI4; Protease inhibitor 4 (kallistatin); Protease inhibitor 4; Serine (or cysteine) proteinase inhibitor; clade A (alpha 1 antiproteinase; antitrypsin); member 4; Serpin A4; Serpin peptidase inhibitor clade A member 4; Serpin peptidase inhibitor; clade A (alpha 1 antiproteinase; antitrypsin); member 4; SERPINA4; Tissue kallistatin inhibitor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5142