Learn More
Biomatik Corporation Human Sialic acid-binding Ig-like lectin 9(SIGLEC9) ELISA kit
Supplier:  Biomatik Corporation EKC42637
Encompass_Preferred
Sample Type: serum,plasma,tissue homogenates.. Species Reactivity: Homo sapiens (Human). Detection Range: 125 pg/ml-8000 pg/ml. Sensitivity: 31.25 pg/ml. Format: 96-Well Plate. Assay Type: Not Available. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9Y336. Target Synonyms: CD329, CDw329, OBBP-LIKE, OB binding protein-like. Restriction: For Research Use Only.
Catalog No. 50-229-5149